N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine 98%

Supplier: Thermo Scientific Chemicals
Danger

H50291-03 H50291-MD
CAAAAH50291-03EA 393.42 CAD
CAAAAH50291-03 CAAAAH50291-MD
N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine 98%
N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine

Purity: 98% CAS No.: 6342-21-8 Molecular Formula: C10H16N2 Formula Weight: 164.25 UN No.: 2735 MDL No.: MFCD01631934

Formula: C₁₀H₁₆N₂
Storage Temperature: Ambient
MDL Number: MFCD01631934
CAS Number: 6342-21-8
UN: 2735
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR