1-(4-Chlorophenyl)cyclopropanecarboxylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50274-03
H50274-06
CAAAAH50274-03EA
76.79
CAD
CAAAAH50274-03
CAAAAH50274-06
1-(4-Chlorophenyl)cyclopropanecarboxylic acid 99%
1-(4-Chlorophenyl)cyclopropanecarboxylic acid
Purity: 99%
CAS No.: 72934-37-3
Molecular Formula: C10H9ClO2
Formula Weight: 196.64
Melting Point: 152-155°
MDL No.: MFCD00001289
Formula:
C₁₀H₉ClO₂ Melting Pt: 152…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001289 CAS Number: 72934-37-3 EINECS: 277-105-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...