4-(Methylsulfonyl)benzenesulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

H50264-03 H50264-06
CAAAAH50264-03EA 100.49 CAD
CAAAAH50264-03 CAAAAH50264-06
4-(Methylsulfonyl)benzenesulfonyl chloride 95%

Purity: 95% CAS No.: 82964-91-8 Molecular Formula: C7H7ClO4S2 Formula Weight: 254.71 Melting Point: 158-163° UN No.: 3261 MDL No.: MFCD00216486

Formula: C₇H₇ClO₄S₂
Melting Pt: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00216486
CAS Number: 82964-91-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR