1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H50263-03
H50263-06
CAAAAH50263-03EA
170.64
CAD
CAAAAH50263-03
CAAAAH50263-06
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane 98%
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)-1,4-diazepane
Purity: 98%
CAS No.: 231953-40-5
Molecular Formula: C11H13ClF3N3
Formula Weight: 279.69
Boiling Point: 128°/1mm
UN No.: 2735
MDL No.: MFCD00238856
Formula:
C₁₁H₁₃ClF₃N₃ Boiling Pt: 128 °C/1 mm Storage Temperature: Ambient |
MDL Number:
MFCD00238856 CAS Number: 231953-40-5 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...