5-Chlorobenzofuran-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chlorobenzo[b]furan-2-carboxylic acid
H50239-03
H50239-06
CAAAAH50239-03EA
161.16
CAD
CAAAAH50239-03
CAAAAH50239-06
5-Chlorobenzofuran-2-carboxylic acid 97%
5-Chlorobenzofuran-2-carboxylic acid
Purity: 97%
CAS No.: 10242-10-1
Molecular Formula: C9H5ClO3
Formula Weight: 196.59
MDL No.: MFCD00060512
Formula:
C₉H₅ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00060512 CAS Number: 10242-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...