2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H50236-03 H50236-MD
CAAAAH50236-03EA 237 CAD
CAAAAH50236-03 CAAAAH50236-MD
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid 98%
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid

Purity: 98% CAS No.: 123533-41-5 Molecular Formula: C14H10N2O2 Formula Weight: 238.25 MDL No.: MFCD00269226

Formula: C₁₄H₁₀N₂O₂
MW: 238.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269226
CAS Number: 123533-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR