4-(Methylsulfonyl)benzoic acid 98%
Supplier: Thermo Scientific Chemicals
H50230-03
H50230-06
CAAAAH50230-03EA
67.31
CAD
CAAAAH50230-03
CAAAAH50230-06
4-(Methylsulfonyl)benzoic acid 98%
4-(Methylsulphonyl)benzoic acid
Purity: 98%
CAS No.: 4052-30-6
Molecular Formula: C8H8O4S
Formula Weight: 200.21
Melting Point: 270-275°
MDL No.: MFCD00007564
Formula:
C₈H₈O₄S Melting Pt: 270…275 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007564 CAS Number: 4052-30-6 EINECS: 223-765-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...