1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H50225-03 H50225-06
CAAAAH50225-03EA 141.25 CAD
CAAAAH50225-03 CAAAAH50225-06
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 98%
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde

Purity: 99% CAS No.: 2644-93-1 Molecular Formula: C7H10N2O Formula Weight: 138.17 Melting Point: 80-83° Boiling Point: 132°/12mm MDL No.: MFCD00159623

Formula: C₇H₁₀N₂O
Boiling Pt: 132 °C/12 mm
Melting Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00159623
CAS Number: 2644-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR