N-Boc-S-methyl-L-cysteine 96%

Supplier: Thermo Scientific Chemicals
Warning

H50217-03 H50217-06
CAAAAH50217-03EA 150.1 CAD
CAAAAH50217-03 CAAAAH50217-06
N-Boc-S-methyl-L-cysteine 96%
N-Boc-S-methyl-L-cysteine

Purity: 96% CAS No.: 16947-80-1 Molecular Formula: C9H17NO4S Formula Weight: 235.30 MDL No.: MFCD00076923

Formula: C₉H₁₇NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00076923
CAS Number: 16947-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR