Boc-Cys(Et)-OH

Supplier: Thermo Scientific Chemicals
Warning

H50216-03 H50216-06
CAAAAH50216-03EA 138.57 CAD
CAAAAH50216-03 CAAAAH50216-06
Boc-Cys(Et)-OH
Boc-Cys(Et)-OH

CAS No.: 16947-82-3 Molecular Formula: C10H19NO4S Formula Weight: 249.33 MDL No.: MFCD00076919

Formula: C₁₀H₁₉NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00076919
CAS Number: 16947-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR