Ac-β-Ala-OH 97%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine

H50208-03 H50208-06
CAAAAH50208-03EA 64.31 CAD
CAAAAH50208-03 CAAAAH50208-06
Ac-β-Ala-OH 97%
Ac-β-Ala-OH

Purity: 97% CAS No.: 3025-95-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 77° MDL No.: MFCD00037296

Formula: C₅H₉NO₃
Melting Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00037296
CAS Number: 3025-95-4
EINECS: 221-185-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR