2-(2-Pyrrolidinyl)pyridine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Pyrrolidin-2-ylpyridine

H50176-03 H50176-MD
CAAAAH50176-03EA 404.48 CAD
CAAAAH50176-03 CAAAAH50176-MD
2-(2-Pyrrolidinyl)pyridine 96%
2-(2-Pyrrolidinyl)pyridine

Purity: 96% CAS No.: 77790-61-5 Molecular Formula: C9H12N2 Formula Weight: 148.21 UN No.: 2810 MDL No.: MFCD01691538

Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01691538
CAS Number: 77790-61-5
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR