Thieno[3,2-b]pyridin-7-ol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
7-Hydroxythieno[3,2-b]pyridine
H50172-03
H50172-06
CAAAAH50172-03EA
83.9
CAD
CAAAAH50172-03
CAAAAH50172-06
Thieno[3,2-b]pyridin-7-ol 96%
Thieno[3,2-b]pyridin-7-ol
Purity: 96%
CAS No.: 107818-20-2
Molecular Formula: C7H5NOS
Formula Weight: 151.19
MDL No.: MFCD01863636
Formula:
C₇H₅NOS MW: 151.183 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863636 CAS Number: 107818-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...