2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

H50139-03 H50139-MD
CAAAAH50139-03EA 206.98 CAD
CAAAAH50139-03 CAAAAH50139-MD
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester 97%
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester

Purity: 97% CAS No.: 947191-69-7 Molecular Formula: C15H22BNO3 Formula Weight: 275.16 MDL No.: MFCD07781184

Formula: C₁₅H₂₂BNO₃
Storage Temperature: Ambient
MDL Number: MFCD07781184
CAS Number: 947191-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR