2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50091-03
H50091-06
CAAAAH50091-03EA
162.74
CAD
CAAAAH50091-03
CAAAAH50091-06
2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde 97%
2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde
Purity: 97%
Molecular Formula: C10H11NO2
Formula Weight: 177.20
MDL No.: MFCD07781136
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...