2-Cyclopropylmethoxy-3-iodopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50084-03
H50084-06
CAAAAH50084-03EA
169.06
CAD
CAAAAH50084-03
CAAAAH50084-06
2-Cyclopropylmethoxy-3-iodopyridine 97%
2-Cyclopropylmethoxy-3-iodopyridine
Purity: 97%
CAS No.: 766557-62-4
Molecular Formula: C9H10INO
Formula Weight: 275.09
MDL No.: MFCD06659010
Formula:
C₉H₁₀INO Storage Temperature: Ambient |
MDL Number:
MFCD06659010 CAS Number: 766557-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...