1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyrrolidine-3-carboxylic acid, N-BOC protected
H50080-03
H50080-MD
CAAAAH50080-03EA
339.7
CAD
CAAAAH50080-03
CAAAAH50080-MD
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 99%
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
Purity: 99%
CAS No.: 59378-75-5
Molecular Formula: C10H17NO4
Formula Weight: 215.25
Melting Point: 128-137°
MDL No.: MFCD02179181
Formula:
C₁₀H₁₇NO₄ MW: 215.25 g/mol Melting Pt: 128…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179181 CAS Number: 59378-75-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...