5-Aminomethyl-2-(boc-amino)pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-(aminomethyl)pyridine, 2-BOC protected, 2-(Boc-amino)-5-(aminomethyl)pyridine

H50078-03 H50078-06
CAAAAH50078-03EA 243.32 CAD
CAAAAH50078-03 CAAAAH50078-06
5-Aminomethyl-2-(boc-amino)pyridine 97%
5-Aminomethyl-2-(boc-amino)pyridine

Purity: 97% CAS No.: 187237-37-2 Molecular Formula: C11H17N3O2 Formula Weight: 223.28 Melting Point: 132-134° MDL No.: MFCD07781132

Formula: C₁₁H₁₇N₃O₂
MW: 223.28 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD07781132
CAS Number: 187237-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR