Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H50074-03 H50074-MD
CAAAAH50074-03EA 317.58 CAD
CAAAAH50074-03 CAAAAH50074-MD
Pyrazolo[1,5-a]pyridine-7-carbaldehyde 97%
Pyrazolo[1,5-a]pyridine-7-carbaldehyde

Purity: 97% CAS No.: 362661-83-4 Molecular Formula: C8H6N2O Formula Weight: 146.15 Melting Point: 83-86° MDL No.: MFCD06739029

Formula: C₈H₆N₂O
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD06739029
CAS Number: 362661-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR