7-Azaindole-5-boronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50044-03
H50044-MD
CAAAAH50044-03EA
206.98
CAD
CAAAAH50044-03
CAAAAH50044-MD
7-Azaindole-5-boronic acid pinacol ester 97%
7-Azaindole-5-boronic acid pinacol ester
Purity: 97%
CAS No.: 754214-56-7
Molecular Formula: C13H17BN2O2
Formula Weight: 244.10
Melting Point: 241-245°
MDL No.: MFCD08060937
Formula:
C₁₃H₁₇BN₂O₂ Melting Pt: 241…245 °C Storage Temperature: Ambient |
MDL Number:
MFCD08060937 CAS Number: 754214-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...