6-Bromo[1,8]naphthyridin-2(1H)-one 98%

Supplier: Thermo Scientific Chemicals
Warning

H50042-03 H50042-MD
CAAAAH50042-03EA 540.36 CAD
CAAAAH50042-03 CAAAAH50042-MD
6-Bromo[1,8]naphthyridin-2(1H)-one 98%
6-Bromo[1,8]naphthyridin-2(1H)-one

Purity: 98% CAS No.: 72754-05-3 Molecular Formula: C8H5BrN2O Formula Weight: 225.05 Melting Point: 300° MDL No.: MFCD07781151

Formula: C₈H₅BrN₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD11226504
CAS Number: 72754-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR