(4-Aminopyridin-3-yl)methanol 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Amino-3-pyridinemethanol, (4-Amino-pyridin-3-yl)-methanol
H50033-03
H50033-06
CAAAAH50033-03EA
229.1
CAD
CAAAAH50033-03
CAAAAH50033-06
(4-Aminopyridin-3-yl)methanol 95%
(4-Aminopyridin-3-yl)methanol
Purity: 97%
CAS No.: 138116-34-4
Molecular Formula: C6H8N2O
Formula Weight: 124.14
Storage Temperature: 2°C to 8°C
Melting Point: 167-169°
MDL No.: MFCD06203066
Formula:
C₆H₈N₂O MW: 124.14 g/mol Melting Pt: 167…169 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06203066 CAS Number: 138116-34-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...