4-Fluoropiperidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

H25755-03 H25755-MD
CAAAH25755-03EA 508.76 CAD
CAAAH25755-03 CAAAH25755-MD
4-Fluoropiperidine hydrochloride 95%
4-Fluoropiperidine hydrochloride

Purity: 95% CAS No.: 57395-89-8 Molecular Formula: C5H10FN.HCl Formula Weight: 139.60 Melting Point: 161-163° MDL No.: MFCD03452786

Formula: C₅H₁₁ClFN
MW: 139.6 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD03452786
CAS Number: 57395-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR