2,2-Difluorocyclopropanecarboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H25752-03
H25752-06
CAAAAH25752-03EA
382.36
CAD
CAAAAH25752-03
CAAAAH25752-06
CAAAAH25752-MD
2,2-Difluorocyclopropanecarboxylic acid 95%
2,2-Difluorocyclopropanecarboxylic acid
Purity: 95%
CAS No.: 107873-03-0
Molecular Formula: C4H4F2O2
Formula Weight: 122.07
Melting Point: 55-60°
Boiling Point: 70°/7mm
UN No.: 3261
MDL No.: MFCD04115828
Formula:
C₄H₄F₂O₂ Boiling Pt: 70 °C/7 mm Melting Pt: 55…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115828 CAS Number: 107873-03-0 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...