3,3-Difluoroazetidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Difluoroazetidine HCl

H25751-03 H25751-MD
CAAAH25751-03EA 734.7 CAD
CAAAH25751-03 CAAAH25751-MD
3,3-Difluoroazetidine hydrochloride 95%
3,3-Difluoroazetidine hydrochloride

Purity: 95% CAS No.: 288315-03-7 Molecular Formula: C3H5F2N.HCl Formula Weight: 129.54 Melting Point: 138-140° MDL No.: MFCD05663714

Formula: C3H6ClF2N
MW: 129.54 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD05663714
CAS Number: 288315-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR