Methyl-3-amino-4-iodobenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-amino-4-iodobenzoate
B25699-03
B25699-06
CAAAAB25699-03EA
87.37
CAD
CAAAAB25699-03
CAAAAB25699-06
Methyl-3-amino-4-iodobenzoate 97%
Methyl 3-amino-4-iodobenzoate
Purity: 97%
CAS No.: 412947-54-7
Molecular Formula: C8H8INO2
Formula Weight: 277.06
Melting Point: 134-136°
MDL No.: MFCD08064225
Formula:
C₈H₈INO₂ MW: 277.06 g/mol Melting Pt: 134…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD08064225 CAS Number: 412947-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...