2,6-Diacetylpyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

B25678-03 B25678-06 B25678-14
CAAAAB25678-03EA 89.9 CAD
CAAAAB25678-03 CAAAAB25678-06 CAAAAB25678-14
2,6-Diacetylpyridine 99%
2,6-Diacetylpyridine

Purity: 99% CAS No.: 1129-30-2 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: 78-82° MDL No.: MFCD00006304

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00006304
CAS Number: 1129-30-2
EINECS: 214-442-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR