1-(6-Methoxy-2-naphthyl)ethanol 98%
Supplier: Thermo Scientific Chemicals
B25628-06
B25628-14
CAAAAB25628-06EA
107.44
CAD
CAAAAB25628-06
CAAAAB25628-14
1-(6-Methoxy-2-naphthyl)ethanol 98%
1-(6-Methoxy-2-naphthyl)ethanol
Purity: 98%
CAS No.: 77301-42-9
Molecular Formula: C13H14O2
Formula Weight: 202.25
Melting Point: 111-112°
MDL No.: MFCD01632581
Formula:
C₁₃H₁₄O₂ Melting Pt: 111…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632581 CAS Number: 77301-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...