(2-Carboxyethyl)triphenylphosphonium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25609-06
B25609-14
CAAAAB25609-06EA
96.06
CAD
CAAAAB25609-06
CAAAAB25609-14
CAAAAB25609-22
(2-Carboxyethyl)triphenylphosphonium chloride 98%
(2-Carboxyethyl)triphenylphosphonium chloride
Purity: 98%
CAS No.: 36626-29-6
Molecular Formula: HO2CCH2CH2P(C6H5)3Cl
Formula Weight: 370.81
Melting Point: 197-201°
MDL No.: MFCD00185663
Formula:
C₂₁H₂₀ClO₂P Melting Pt: 197…201 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185663 CAS Number: 36626-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...