3,3-Dimethyl-2-oxobutyric acid ∼60% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

B25582-18 B25582-30
CAAAAB25582-18EA 61.46 CAD
CAAAAB25582-18 CAAAAB25582-30
3,3-Dimethyl-2-oxobutyric acid ∼60% in aqueous solution
3,3-Dimethyl-2-oxobutyric acid

CAS No.: 815-17-8 Molecular Formula: C6H10O3 Formula Weight: 130.14 UN No.: 3265 MDL No.: MFCD00154352

Formula: C₆H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00154352
CAS Number: 815-17-8
EINECS: 212-418-2
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR