N-(Diphenylmethylene)glycine ethyl ester 98%

Supplier: Thermo Scientific Chemicals

B25580-06 B25580-14
CAAAAB25580-06EA 122.45 CAD
CAAAAB25580-06 CAAAAB25580-14
N-(Diphenylmethylene)glycine ethyl ester 98%
N-(Diphenylmethylene)glycine ethyl ester

Purity: 98% CAS No.: 69555-14-2 Molecular Formula: C17H17NO2 Formula Weight: 267.33 Flash Point: 110°(230°F) Melting Point: 51-55° MDL No.: MFCD00010590

Formula: C₁₇H₁₇NO₂
Melting Pt: 51…55 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010590
CAS Number: 69555-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR