Malonamamidine hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Malonamideamidine hydrochloride

B25569-14 B25569-22
CAAAAB25569-14EA 134.77 CAD
CAAAAB25569-14 CAAAAB25569-22
Malonamamidine hydrochloride ≥98%
Malonamamidine hydrochloride

Purity: 98+% CAS No.: 34570-17-7 Molecular Formula: C3H7N3O.HCl Formula Weight: 137.57 Melting Point: 170-174° MDL No.: MFCD00013007

Formula: C₃H₇N₃O·HCl
MW: 137.57 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00013007
CAS Number: 34570-17-7
EINECS: 252-095-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR