Methyl-1-cyclohexene-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl cyclohex-1-enecarboxylate

B25568-06 B25568-14
CAAAAB25568-06EA 210.14 CAD
CAAAAB25568-06 CAAAAB25568-14
Methyl-1-cyclohexene-1-carboxylate 97%
Methyl-1-cyclohexene-1-carboxylate

Purity: 97% CAS No.: 18448-47-0 Molecular Formula: C8H12O2 Formula Weight: 140.18 Flash Point: 73°(165°F) Refractive Index: 1.4770 Boiling Point: 190-192° MDL No.: MFCD00001544

Formula: C₈H₁₂O₂
MW: 140.182 g/mol
Boiling Pt: 190…192 °C
Density: 1.030
Flash Pt: 73 °C(165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001544
CAS Number: 18448-47-0
EINECS: 242-331-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR