Methyl (4-fluorobenzoyl)acetate 95%

Supplier: Thermo Scientific Chemicals

B25546-03 B25546-09
CAAAAB25546-03EA 77.26 CAD
CAAAAB25546-03 CAAAAB25546-09
Methyl (4-fluorobenzoyl)acetate 95%
Methyl (4-fluorobenzoyl)acetate

Purity: 95% CAS No.: 63131-29-3 Molecular Formula: C10H9FO3 Formula Weight: 196.18 Flash Point: 110°(230°F) Refractive Index: 1.5210 Boiling Point: 91°/5mm MDL No.: MFCD00000355

Formula: C₁₀H₉FO₃
Boiling Pt: 91 °C/5 mm
Density: 1.228
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000355
CAS Number: 63131-29-3
EINECS: 263-889-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR