(±)-Ethyl-2-bromovalerate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-bromovalerate
B25530-14
B25530-22
CAAAAB25530-14EA
49.77
CAD
CAAAAB25530-14
CAAAAB25530-22
(±)-Ethyl-2-bromovalerate 97%
(±)-Ethyl 2-bromovalerate
Purity: 97%
CAS No.: 615-83-8
Molecular Formula: C7H13BrO2
Formula Weight: 209.09
Flash Point: 77°(170°F)
Refractive Index: 1.4480
Boiling Point: 190-192°
Beilstein Reference: 1098901
UN No.: 3265
MDL No.: MFCD00000159
Formula:
C₇H₁₃BrO₂ MW: 209.09 g/mol Boiling Pt: 190…192 °C Density: 1.226 Flash Pt: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000159 CAS Number: 615-83-8 EINECS: 210-450-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...