Coumalic acid 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Oxo-2H-pyran-5-carboxylic acid

B25521-06 B25521-14
CAAAAB25521-06EA 40.29 CAD
CAAAAB25521-06 CAAAAB25521-14
Coumalic acid 97%
Coumalic acid

Purity: 97% CAS No.: 500-05-0 Molecular Formula: C6H4O4 Formula Weight: 140.10 Melting Point: ca 208° dec. Boiling Point: 218°/120mm Merck Reference: 14,2557 MDL No.: MFCD00006644

Formula: C₆H₄O₄
MW: 140.10 g/mol
Boiling Pt: 218 °C/120 mm
Melting Pt: ∼ 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006644
CAS Number: 500-05-0
EINECS: 207-899-0
Merck Index: 14,2557

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR