4'-(4-Methylphenyl)-2,2':6',2''-terpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(p-Tolyl)-2,2:6,2-terpyridine

B25509-02 B25509-77
CAAAAB25509-02EA 254.38 CAD
CAAAAB25509-02 CAAAAB25509-77
4'-(4-Methylphenyl)-2,2':6',2''-terpyridine 98%
4'-(4-Methylphenyl)-2,2':6',2''-terpyridine

Purity: 98% CAS No.: 89972-77-0 Molecular Formula: C22H17N3 Formula Weight: 323.40 Melting Point: 167-170° MDL No.: MFCD06796977

Formula: C₂₂H₁₇N₃
MW: 323.40 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD06796977
CAS Number: 89972-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR