5-Bromo-2-hydroxybenzophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25507-06
B25507-14
CAAAAB25507-06EA
133.83
CAD
CAAAAB25507-06
CAAAAB25507-14
5-Bromo-2-hydroxybenzophenone 97%
5-Bromo-2-hydroxybenzophenone
Purity: 97%
CAS No.: 55082-33-2
Molecular Formula: C13H9BrO2
Formula Weight: 277.11
Melting Point: 108-111°
MDL No.: MFCD00525062
Formula:
C₁₃H₉BrO₂ Melting Pt: 108…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00525062 CAS Number: 55082-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...