5-Methylisophthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B25496-03 B25496-06
CAAAAB25496-03EA 36.66 CAD
CAAAAB25496-03 CAAAAB25496-06
5-Methylisophthalic acid 98%
5-Methylisophthalic acid

Purity: 98% CAS No.: 499-49-0 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 296-300° MDL No.: MFCD00013986

Formula: C₉H₈O₄
Melting Pt: 296…300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013986
CAS Number: 499-49-0
EINECS: 207-881-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR