5-(Trifluoromethoxy)isatin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-(Trifluoromethoxy)indoline-2,3-dione

B25494-03 B25494-06
CAAAAB25494-03EA 79.63 CAD
CAAAAB25494-03 CAAAAB25494-06
5-(Trifluoromethoxy)isatin 98%
5-(Trifluoromethoxy)isatin

Purity: 98% CAS No.: 169037-23-4 Molecular Formula: C9H4F3NO2 Formula Weight: 231.13 Melting Point: 170-172° MDL No.: MFCD00192524

Formula: C₉H₄F₃NO₃
MW: 231.13 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00192524
CAS Number: 169037-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR