4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25486-02
B25486-77
CAAAAB25486-02EA
108.39
CAD
CAAAAB25486-02
CAAAAB25486-77
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine 98%
4'-(4-Methoxyphenyl)-2,2':6',2''-terpyridine
Purity: 98%
CAS No.: 13104-56-8
Molecular Formula: C22H17N3O
Formula Weight: 339.40
Melting Point: 161-163°
MDL No.: MFCD06796987
Formula:
C₂₂H₁₇N₃O Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD06796987 CAS Number: 13104-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...