5-Chloro-2,4-dinitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25479-06
B25479-14
CAAAAB25479-06EA
199.08
CAD
CAAAAB25479-06
CAAAAB25479-14
5-Chloro-2,4-dinitrobenzoic acid 98%
5-Chloro-2,4-dinitrobenzoic acid
Purity: 98%
CAS No.: 136833-36-8
Molecular Formula: C7H3ClN2O6
Formula Weight: 246.56
Melting Point: 180-182°
MDL No.: MFCD00091829
Formula:
C₇H₃ClN₂O₆ Melting Pt: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00091829 CAS Number: 136833-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...