(±)-Dimethyl acetylsuccinate 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Dimethyl acetylsuccinate

B25423-22 B25423-36
CAAAAB25423-22EA 78.68 CAD
CAAAAB25423-22 CAAAAB25423-36
(±)-Dimethyl acetylsuccinate 96%
(±)-Dimethyl acetylsuccinate

Purity: 96% CAS No.: 10420-33-4 Molecular Formula: C8H12O5 Formula Weight: 188.18 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 129-134°/12mm MDL No.: MFCD00008448

Formula: C₈H₁₂O₅
MW: 188.18 g/mol
Boiling Pt: 129…134 °C/12 mm
Melting Pt: 30…33 °C
Density: 1.160
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008448
CAS Number: 10420-33-4
EINECS: 233-897-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR