Ethyl-4-pyrazolecarboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
ethyl 1H-pyrazole-4-carboxylate, Ethyl 4-pyrazolecarboxylate
B25351-06
B25351-14
CAAAAB25351-03EA
86.2
CAD
CAAAAB25351-03
CAAAAB25351-06
CAAAAB25351-14
Ethyl-4-pyrazolecarboxylate 98%
Ethyl-4-pyrazolecarboxylate
Purity: 98%
CAS No.: 37622-90-5
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Melting Point: 75-80°
Boiling Point: 138-140°/3mm
MDL No.: MFCD00010844
Formula:
C₆H₈N₂O₂ Boiling Pt: 138…140 °C/3 mm Melting Pt: 75…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010844 CAS Number: 37622-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...