Aniline-2,5-disulfonic acid 95%

Supplier: Thermo Scientific Chemicals

B25350-06 B25350-14
CAAAAB25350-06EA 104.44 CAD
CAAAAB25350-06 CAAAAB25350-14
Aniline-2,5-disulfonic acid 95%
Aniline-2,5-disulphonic acid

Purity: 95% CAS No.: 98-44-2 Molecular Formula: C6H7NO6S2 Formula Weight: 253.25 Melting Point: 300° MDL No.: MFCD00035753

Formula: C₆H₇NO₆S₂
MW: 253.25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00035753
CAS Number: 98-44-2
EINECS: 202-669-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR