Salicylideneaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25332-09
B25332-18
CAAAAB25332-09EA
102.38
CAD
CAAAAB25332-09
CAAAAB25332-18
Salicylideneaniline 98%
Salicylideneaniline
Purity: 98%
CAS No.: 779-84-0
Molecular Formula: C13H11NO
Formula Weight: 197.24
Melting Point: 48-52°
Boiling Point: 151°/2mm
MDL No.: MFCD00020066
Formula:
C₁₃H₁₁NO Boiling Pt: 151 °C/2 mm Melting Pt: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020066 CAS Number: 779-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...