Methyl coumalate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-oxo-2H-pyran-5-carboxylate

B25330-04 B25330-09
CAAAAB25330-04EA 88.32 CAD
CAAAAB25330-04 CAAAAB25330-09
Methyl coumalate 98%
Methyl coumalate

Purity: 98% CAS No.: 6018-41-3 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: 68-72° Boiling Point: 178-180°/60mm Merck Reference: 14,2557 MDL No.: MFCD00010120

Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 178…180 °C/60 mm
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00010120
CAS Number: 6018-41-3
EINECS: 227-871-1
Merck Index: 14,2557

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR