Methyl malonamate 98%
Supplier: Thermo Scientific Chemicals
B25326-14
B25326-22
CAAAAB25326-14EA
693.62
CAD
CAAAAB25326-14
CAAAAB25326-22
Methyl malonamate 98%
Methyl malonamate
Purity: 98%
CAS No.: 51513-29-2
Molecular Formula: C4H7NO3
Formula Weight: 117.11
Melting Point: 38-40°
MDL No.: MFCD00674533
Formula:
C₄H₇NO₃ Melting Pt: 38…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674533 CAS Number: 51513-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...