2-Phenylbenzimidazole-5-sulfonic acid monohydrate 98%

Supplier: Thermo Scientific Chemicals

B25307-06 B25307-14
CAAAAB25307-06EA 58.46 CAD
CAAAAB25307-06 CAAAAB25307-14
2-Phenylbenzimidazole-5-sulfonic acid monohydrate 98%
2-Phenylbenzimidazole-5-sulfonic acid monohydrate

Purity: 98% CAS No.: 27503-81-7 Molecular Formula: C13H10N2O3S.H2O Formula Weight: 292.32 (274.30anhy) Melting Point: 300° Beilstein Reference: 922664 MDL No.: MFCD00150559

Formula: C₁₃H₁₀N₂O₃S·H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150559
CAS Number: 27503-81-7
EINECS: 248-502-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR