3-Amino-4-chloro-N,N-dimethylbenzamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25294-06
B25294-14
B25294-22
CAAAAB25294-06EA
38.39
CAD
CAAAAB25294-06
CAAAAB25294-14
CAAAAB25294-22
3-Amino-4-chloro-N,N-dimethylbenzamide 97%
3-Amino-4-chloro-N,N-dimethylbenzamide
Purity: 97%
CAS No.: 19694-10-1
Molecular Formula: C7H7ClN2O
Formula Weight: 170.60
Melting Point: 164-166°
MDL No.: MFCD00035785
Formula:
C₇H₇ClN₂O Melting Pt: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035785 CAS Number: 19694-10-1 EINECS: 243-231-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...